Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF165 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541101
Description
Peptide sequence: ISEASGESQD ICKSAGRVKR QWEKESGESQ RLSSAQDEGF GKILTHKNTV Sequence homology: Cow: 79%; Human: 100%; Pig: 86%.
This protein is a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. The protein may play a role in spermatogenesis.This gene encodes a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. This gene is located within a cluster of zinc finger family members. The encoded protein may play a role in spermatogenesis.
Specifications
| ZNF165 | |
| Polyclonal | |
| Unconjugated | |
| ZNF165 | |
| Cancer/testis antigen 53; CT53; LD65; Unknown (protein for MGC:140342); zinc finger and SCAN domain-containing protein 7; Zinc finger protein 165; ZNF165; ZPF165; ZSCAN7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7718 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P49910 | |
| ZNF165 | |
| Synthetic peptide directed towards the middle region of human ZNF165. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction