Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF165 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582290 Supplier Invitrogen™ Supplier No. PA582290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWVHEHYPES GEEAVTILED LERGTDEAVL QVQAHEHGQE IFQKKVSPPG PALNVKLQPV ETKAHFDSSE PQLLWDCDNE SENSRSMPKL EIFEKIESQR IISGRISGYI SEASGESQ Highest antigen sequence indentity to the following orthologs: Mouse - 32%, Rat - 31%.

This protein is a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. The protein may play a role in spermatogenesis.This gene encodes a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. This gene is located within a cluster of zinc finger family members. The encoded protein may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF165
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF165
Gene Accession No. P49910
Gene Alias Cancer/testis antigen 53; CT53; LD65; Unknown (protein for MGC:140342); zinc finger and SCAN domain-containing protein 7; Zinc finger protein 165; ZNF165; ZPF165; ZSCAN7
Gene Symbols ZNF165
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF165. Recombinant protein control fragment (Product #RP-89440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.