Learn More
Invitrogen™ ZNF167 Monoclonal Antibody (3A12)

Description
Peptide Sequence: TTKESPPTSP LSGGSAPGAH LEPPYDPGTH HLPSGDFAQC TSPVPTLPQV GNSGDQAGAT VLRMVRPQDT VAYEDLSVDY TQKKWKSLTL SQRALQWNMM.
Specifications
Specifications
| Antigen | ZNF167 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3A12 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZKSCAN7 |
| Gene Accession No. | Q9P0L1 |
| Gene Alias | ZFP; Zfp167; Zinc finger protein 167; zinc finger protein 448; Zinc finger protein 64; zinc finger protein with KRAB and SCAN domains 7; zinc finger with KRAB and SCAN domains 7; ZKSCAN7; ZNF167; ZNF448; ZNF64; ZSCAN39 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.