Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF167 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562087 Supplier Invitrogen™ Supplier No. PA562087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAQKQEMHFE ETTALGTTKE SPPTSPLSGG SAPGAHLEPP YDPGTHHLPS GDFAQCTSPV PTLPQVGNSG DQAG Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 45%.

ZNF167, a gene located on chromosome 3, encodes for a zinc finger protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF167
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZKSCAN7
Gene Accession No. Q9P0L1
Gene Alias ZFP; Zfp167; Zinc finger protein 167; zinc finger protein 448; Zinc finger protein 64; zinc finger protein with KRAB and SCAN domains 7; zinc finger with KRAB and SCAN domains 7; ZKSCAN7; ZNF167; ZNF448; ZNF64; ZSCAN39
Gene Symbols ZKSCAN7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF167. Recombinant protein control fragment (Product #RP-100850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.