Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF167 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541158 Supplier Invitrogen™ Supplier No. PA541158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGLIPRSTAF QKQEGRLTVK QEPANQTWGQ GSSLQKNYPP VCEIFRLHFR Sequence homology: Human: 100%; Rat: 79%.

ZNF167, a gene located on chromosome 3, encodes for a zinc finger protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF167
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZKSCAN7
Gene Accession No. Q9P0L1
Gene Alias ZFP; Zfp167; Zinc finger protein 167; zinc finger protein 448; Zinc finger protein 64; zinc finger protein with KRAB and SCAN domains 7; zinc finger with KRAB and SCAN domains 7; ZKSCAN7; ZNF167; ZNF448; ZNF64; ZSCAN39
Gene Symbols ZKSCAN7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF167 (aa 9-58).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.