Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590050 Supplier Invitrogen™ Supplier No. PA590050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LASVGYQLCR HSLISKVDQE QLKTDERGIL QGDCADWETQ LKPKDTIAMQ NIPGGKTSNG I; Positive Samples: Mouse testis; Cellular Location: Nucleus.

ZNF177 (Zinc Finger Protein 177) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includenucleic acid binding. An important paralog of this gene isZNF559-ZNF177.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF177
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias PIGX; zinc finger protein 177; ZNF177
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-110 of human ZNF177 (NP_0034422).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.