Learn More
Invitrogen™ ZNF179 Polyclonal Antibody

Description
ZNF179 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ZNF179 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | RNF112 |
| Gene Accession No. | Q96DY5, Q9ULX5 |
| Gene Alias | AI854096; Bfb; bfp; Brain finger protein; LOW QUALITY PROTEIN: RING finger protein 112; neurolastin; R75574; RING finger protein 112; Rnf112; Zfp179; zinc finger protein 179; ZNF179 |
| Gene Symbols | RNF112 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.