Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF179 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553402
Description
Immunogen sequence: LAQEIKNLSG WMGRTGPGFT SPDEMAAQLH DLRKVEAAKR EFEEYVRQQD VATKRIFSAL RVLPDTMRNL LSTQKDAILA RHGVALLCKG RDQTLEALEA ELQATAKAFM DSYT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
ZNF179 | |
Polyclonal | |
Unconjugated | |
RNF112 | |
AI854096; Bfb; bfp; Brain finger protein; LOW QUALITY PROTEIN: RING finger protein 112; neurolastin; R75574; RING finger protein 112; Rnf112; Zfp179; zinc finger protein 179; ZNF179 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULX5 | |
RNF112 | |
Recombinant protein corresponding to Human ZNF179. Recombinant protein control fragment (Product #RP-90951). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction