Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552259 Supplier Invitrogen™ Supplier No. PA552259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISIQVLGQDI LSEKMESPSC QVGEVEPHLE VVPQELGLEN SSSGPGELLS HIVKEESDTE AELALAASQP ARLEERLIRD QDLGASLLPA APQEQWRQLD STQKEQYWDL MLETYGKMVS GAGISHPKSD LTNS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.

ZNF18 be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF18
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF18
Gene Accession No. P17022
Gene Alias 1700128E15Rik; D11Ertd714e; HDSG1; Heart development-specific gene 1 protein; KOX11; Zfp18; Zfp535; Zinc finger protein 18; zinc finger protein 18 (KOX 11); zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6; Zkscan6; Znf18; ZNF535; ZSCAN38
Gene Symbols ZNF18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF18. Recombinant protein control fragment (Product #RP-89371).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7566
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.