Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106471 Supplier Invitrogen™ Supplier No. PA5106471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF18.

ZNF18 be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF18
Gene Accession No. P17022, Q642B9, Q810A1
Gene Alias 1700128E15Rik; D11Ertd714e; HDSG1; Heart development-specific gene 1 protein; KOX11; Zfp18; Zfp535; Zinc finger protein 18; zinc finger protein 18 (KOX 11); zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6; Zkscan6; Znf18; ZNF535; ZSCAN38
Gene Symbols Zfp18, Zkscan6, ZNF18
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF18(Accession P17022), corresponding to amino acid residues D450-S500.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303226, 52712, 7566
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.