Learn More
Invitrogen™ ZNF180 Monoclonal Antibody (4F3)

Description
Peptide Sequence: VAFTQRKAVI HERVCKSDET GEKSGLNSSL FSSPVIPIRN HFHKHVSHAK KWHLNAAVNS HQKINENETL YENNECGKPP QSIHLIQFTR TQTKDKSYGF SDRIQSFCHG.
Specifications
Specifications
| Antigen | ZNF180 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4F3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF180 |
| Gene Accession No. | Q9UJW8 |
| Gene Alias | HHZ168; zinc finger protein 180; ZNF180 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.