Learn More
Invitrogen™ ZNF181 Monoclonal Antibody (5F1)

Description
Peptide Sequence: FIHRSSLIHH QKIHTGEKPY ECRECGKAFC CSSHLTRHQR IHTMEKQYEC NKCLKVFSSL SFLVQHQSIH TEEKPFECQK CRKSFNQLES LNMHLRNHIR LKPYECSI.
Specifications
Specifications
| Antigen | ZNF181 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5F1 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF181 |
| Gene Accession No. | Q2M3W8 |
| Gene Alias | HHZ181; Zinc finger protein 181; ZNF181 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.