Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF181 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584546 Supplier Invitrogen™ Supplier No. PA584546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHNGEKPNSV VSVEKPLDHM NPYTCEKSYR RET Highest antigen sequence indentity to the following orthologs: Mouse - 42%, Rat - 38%.

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF181
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF181
Gene Accession No. Q2M3W8
Gene Alias HHZ181; Zinc finger protein 181; ZNF181
Gene Symbols ZNF181
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF181. Recombinant protein control fragment (Product #RP-107157).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.