Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF182 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220969
Description
ZNF182 Polyclonal specifically detects ZNF182 in Human samples. It is validated for Western Blot.Specifications
ZNF182 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
HHZ150, KOX14zinc finger protein 182 (HHZ150), MGC131713, Zfp182, zinc finger protein 182, Zinc finger protein 21, zinc finger protein 21 (KOX 14), Zinc finger protein KOX14, ZNF21MGC125383 | |
Rabbit | |
74 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
P17025 | |
ZNF182 | |
Recombinant protein encompassing a sequence within the N-terminus region of human ZNF182. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
7569 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction