Learn More
Invitrogen™ ZNF182 Monoclonal Antibody (6D11)

Description
Peptide Sequence: VEECPAEGKI PFWNFPEVCQ VDEQIERQHQ DDQDKCLLMQ VGFSDKKTII TKSARDCHEF GNILHLSTNL VASIQRPDKH ESFGNNMVDN LDLFSRSSAE NKYDNGCAKL.
Specifications
Specifications
| Antigen | ZNF182 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6D11 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF182 |
| Gene Accession No. | P17025 |
| Gene Alias | 6330583I20Rik; HHZ150; KOX14; MGC137263 protein; Zfp182; Zinc finger protein 182; zinc finger protein 21; zinc finger protein 21 (KOX 14); zinc finger protein KOX14; ZNF182; ZNF21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.