Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF182 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584425 Supplier Invitrogen™ Supplier No. PA584425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSNLVFVGQQ VTKPNLILKL EVEECPAEGK IPFWNFPEVC QVDE Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 51%.

Zinc-finger proteins bind nucleic acids and play important roles in various cellular functions, including cell proliferation, differentiation, and apoptosis. This gene encodes a zinc finger protein, and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF182
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF182
Gene Accession No. P17025
Gene Alias 6330583I20Rik; HHZ150; KOX14; MGC137263 protein; Zfp182; Zinc finger protein 182; zinc finger protein 21; zinc finger protein 21 (KOX 14); zinc finger protein KOX14; ZNF182; ZNF21
Gene Symbols ZNF182
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF182. Recombinant protein control fragment (Product #RP-101936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7569
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.