Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF184 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551866 Supplier Invitrogen™ Supplier No. PA551866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSKPDVISQL EQGTEPWIME PSIPVGTCAD WETRLENSVS APEPDISEEE LSPEVIVEKH KRDDSWSSNL LESWEYEGSL ERQQANQQTL PKEIKVTEKT IPSWEKGPVN NEFGKSVNVS SNLVTQEPSP EETSTKRSIK QN Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 49%.

The protein encoded by this gene is predicted to be a Kruppel C2H2-type zinc-finger protein family member. Sequence analysis predicts that the protein contains two Kruppel associated box (KRAB) boxes in the N-terminus and highly conserved zinc finger motifs at the C-terminus. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF184
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF184
Gene Accession No. Q99676
Gene Alias kr-ZNF3; Zinc finger protein 184; zinc finger protein 184 (Kruppel-like); ZNF184
Gene Symbols ZNF184
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF184. Recombinant protein control fragment (Product #RP-99737).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.