Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF185 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | ZNF185 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18032020
![]() |
Novus Biologicals
NBP18032020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180320
![]() |
Novus Biologicals
NBP180320 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZNF185 Polyclonal specifically detects ZNF185 in Human samples. It is validated for Western Blot.Specifications
| ZNF185 | |
| Polyclonal | |
| Rabbit | |
| NP_009081 | |
| 7739 | |
| Synthetic peptide directed towards the N terminal of human ZNF185. Peptide sequence LAPYNIRRSSTSGDTEEEEEEEVVPFSSDEQKRRSEAASGVLRRTAPREH. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| zinc finger protein 185 (LIM domain) | |
| ZNF185 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title