Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF187 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220970
Description
ZNF187 Polyclonal specifically detects ZNF187 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNF187 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
MGC2815, Protein SRE-ZBP, Zinc finger and SCAN domain-containing protein 26, zinc finger protein 187, ZSCAN26SRE-ZBP | |
Rabbit | |
55 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Bovine (93%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.77mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q16670 | |
ZNF187 | |
Recombinant protein encompassing a sequence within the center region of human ZNF187. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
7741 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction