Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF189 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557073 Supplier Invitrogen™ Supplier No. PA557073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDEEPTVKQE IEEIEEEVEP QGVIVTRIKS EIDQDPMGRE TFELVGRLDK QRGIFLWEIP RESLTQEQRM FRENTNIIRK RPN Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.

Kruppel-like zinc finger proteins such as ZNF189 contain a conserved stretch of 7 amino acids that connects a variable number of DNA-binding zinc finger repeats of the cys(2)his(2) (C2H2) type (summarized by Odeberg et al., 1998 [PubMed 9653648]). Approximately 30% of human Kruppel-like zinc finger proteins contain an N-terminal Kruppel-associated box (KRAB) domain. The KRAB domain consists of approximately 75 amino acids that may be subdivided into an A box, which is present in every KRAB domain and is essential for transcriptional repression, and a B box, which is not always present.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF189
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF189
Gene Accession No. O75820
Gene Alias Zinc finger protein 189; ZNF189
Gene Symbols ZNF189
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF189. Recombinant protein control fragment (Product #RP-97597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7743
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.