Learn More
Invitrogen™ ZNF192 Monoclonal Antibody (3B10)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA520289
Catalog No. PIMA520289
Only null left
Add to Cart
Description
Description
Peptide Sequence: MAEESRKPSA PSPPDQTPEE DLVIVKVEED HGWDQESSLH ESNPLGQEVF RLRFRQLRYQ ETLGPREALI QLRALCHQWL RPDLNTKEQI.
May be involved in transcriptional regulation. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF192
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q15776
ZKSCAN8
ZKSCAN8 (NP_006289.2, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunohistochemistry (PFA fixed), Western Blot
3B10
Unconjugated
ZKSCAN8
2510038J07Rik; CAML; D430019P06Rik; LD5-1; Zfp192; zinc finger protein 192; Zinc finger protein with KRAB and SCAN domains 8; zinc finger with KRAB and SCAN domains 8; ZKSCAN8; ZNF192; ZSCAN40
Mouse
Affinity chromatography
RUO
7745
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.