Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF197 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541523 Supplier Invitrogen™ Supplier No. PA541523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MTRENVAHNA LRQEGLVKGK DDTWKWGTSF QGSSSSVWET SHLHFRQLRY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Pig: 93%; Rat: 100%.

ZNF197 product belongs to the zinc finger protein superfamily, members of which are regulatory proteins characterized by nucleic acid-binding zinc finger domains. The encoded protein contains 20 tandemly arrayed C2H2-type zinc fingers, a Kruppel-associated box (KRAB) domain, and a SCAN box. This transcript turns over rapidly and contains 3' UTR AUUUA motifs, which are often a hallmark of rapid turnover. It is overexpressed in some thyroid papillary carcinomas. ZNF197 is located in a cluster of zinc finger genes at 3p21. Two alternatively spliced transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF197
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF197
Gene Accession No. O14709
Gene Alias D3S1363E; P18; pVHL-associated KRAB domain-containing protein; VHLaK; VHL-associated KRAB-A domain-containing protein; zinc finger protein 166; zinc finger protein 197; Zinc finger protein 197-like protein; zinc finger protein 20; zinc finger protein with KRAB and SCAN domains 9; ZKSCAN9; ZNF166; ZNF197; ZnF20; ZSCAN41
Gene Symbols ZNF197
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF197 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.