Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530208
Description
Recommended positive controls: HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The ZNF2 gene is part of a sub-family of structurally related finger protein genes. Members of this sub-family are expressed as multiple transcripts in several cell lines.
Specifications
| ZNF2 | |
| Polyclonal | |
| Unconjugated | |
| ZNF2 | |
| 2810405K07Rik; A1-5; Mkr2; Mkr-2; multifinger protein mKr2; Zfp2; Zfp661; zfp-661; zinc finger protein 2; zinc finger protein 2 (A1-5); zinc finger protein 2 homolog; zinc finger protein 2.2; Zinc finger protein 661; ZNF2; ZNF661 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7549 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BSG1 | |
| ZNF2 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction