Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598253
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598253 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598253 Supplier Invitrogen™ Supplier No. PA598253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The ZNF2 gene is part of a sub-family of structurally related finger protein genes. Members of this sub-family are expressed as multiple transcripts in several cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF2
Gene Accession No. Q9BSG1
Gene Alias 2810405K07Rik; A1-5; Mkr2; Mkr-2; multifinger protein mKr2; Zfp2; Zfp661; zfp-661; zinc finger protein 2; zinc finger protein 2 (A1-5); zinc finger protein 2 homolog; zinc finger protein 2.2; Zinc finger protein 661; ZNF2; ZNF661
Gene Symbols ZNF2
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 2 protein (134-289AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7549
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.