Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145044
Description
Immunogen sequence: SYLDSFQSHD KACTKEKPYD GKECTET Antigen sequence identity: Mouse 52%; Rat 52%.
Zinc finger protein 20, May be involved in transcriptional regulation.
Specifications
ZNF20 | |
Polyclonal | |
Unconjugated | |
ZNF20 | |
KOX13; Zinc finger protein 20; zinc finger protein KOX13; ZNF20 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7568 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17024 | |
ZNF20 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 153-179. Recombinant protein control fragment (Product #RP-110257). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction