Learn More
Invitrogen™ ZNF20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121090
Description
Positive test controls include: SKOV3, A-549, Mouse kidney, Mouse lung, Rat kidney. The target is usually found in the following locations: Nucleus. Immunogen sequence: MMFQDSVAFE DVAVSFTQEE WALLDPSQKN LYRDVMQETF KNLTSVGKTW KVQNIEDEYK NPRRNLSLMR EKLCESKESH HCGESFNQIA DDMLNRKTLP GITPCESSVC GEVGTGHSSL NTHIRADTGH KSSEYQEYGE NPYRNKECKK AFSYLDSFQS HDKACTKEKP YDGKECTETF ISHSCIQRHR VMHSGDGPYK.
Zinc finger protein 20, May be involved in transcriptional regulation.
Specifications
ZNF20 | |
Polyclonal | |
Unconjugated | |
ZNF20 | |
KOX13; Zinc finger protein 20; zinc finger protein KOX13; ZNF20 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7568 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17024 | |
ZNF20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ZNF20 (NP_066966.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.