Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF200 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551277
Description
The antibody detects endogenous levels of total ZNF200 protein.
Could have a role in spermatogenesis.
Specifications
ZNF200 | |
Polyclonal | |
Unconjugated | |
ZNF200 | |
MGC45293; Zinc finger protein 200; ZN200; ZNF200; ZNFMF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P98182 | |
ZNF200 | |
Synthetic peptide of human ZNF200. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction