Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF202 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140097 Supplier Invitrogen™ Supplier No. PA5140097
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PWVPDIQEPQ ETQEPEILSF TYTGDRSKDE EECLEQEDLS LEDIHRPVLG EPEIHQTPDW EIVFEDNPGR LNERRFGTNI SQVNS Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 64%.

ZNF202 (Zinc finger protein 202) is a transcriptional repressor of genes affecting the vascular endothelium as well as lipid metabolism and energy homeostasis. Among its targets are structural components of lipoprotein particles (apolipoproteins AIV, CIII, and E), enzymes involved in lipid processing (lipoprotein lipase, lecithin cholesteryl ester transferase), transporters involved in lipid homeostasis (ABCA1, ABCG1), and several genes involved in processes related to energy metabolism and vascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF202
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF202
Gene Accession No. O95125
Gene Alias Zinc finger protein 202; zinc finger protein with KRAB and SCAN domains 10; ZKSCAN10; ZNF202; ZSCAN42
Gene Symbols ZNF202
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF202. Recombinant protein control fragment (Product #RP-108967).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.