Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF205 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306486IR
This item is not returnable.
View return policy
Description
ZNF205 Polyclonal antibody specifically detects ZNF205 in Human samples. It is validated for ImmunofluorescenceSpecifications
ZNF205 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
7755 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 755 | |
zinc finger protein 205, Zinc finger protein 210, ZNF210Zfp13 | |
Produced in rabbits immunized with E. coli-derived Human ZNF205 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction