Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF205 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541462 Supplier Invitrogen™ Supplier No. PA541462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PSKLGEAVPS GDTQESLHIK MEPEEPHSEG ASQEDGAQGA WGWAPLSHGS Sequence homology: Cow: 85%; Dog: 79%; Guinea Pig: 79%; Human: 100%; Mouse: 93%; Rat: 86%.

ZNF205 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF205
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF205
Gene Accession No. O95201
Gene Alias RhitH; transcriptional repressor RhitH; Zfp13; Zinc finger protein 205; zinc finger protein 210; Znf205; ZNF210
Gene Symbols ZNF205
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF205 (aa 31-80).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7755
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.