Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF205 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551636 Supplier Invitrogen™ Supplier No. PA551636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DHTQQNFYRD VLQKKNGLSL GFPFSRPFWA PQAHGKGEAS GSSRQAGDEK EWRGACTGAV EVGQRVQTSS VAALGNVKPF RTRAGRVQWG VPQCAQEAAC GRSSGPAKDS GQPAEPDRTP DAAPPDPSPT EPQEYRVPEK PNEE Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 24%.

ZNF205 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF205
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF205
Gene Accession No. O95201
Gene Alias RhitH; transcriptional repressor RhitH; Zfp13; Zinc finger protein 205; zinc finger protein 210; Znf205; ZNF210
Gene Symbols ZNF205
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF205. Recombinant protein control fragment (Product #RP-99733).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.