Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF205 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107318
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%.
ZNF205 may be involved in transcriptional regulation.
Specifications
ZNF205 | |
Polyclonal | |
Unconjugated | |
ZNF205 | |
RhitH; transcriptional repressor RhitH; Zfp13; Zinc finger protein 205; zinc finger protein 210; Znf205; ZNF210 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7755 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
O95201 | |
ZNF205 | |
Synthetic peptide directed towards the middle region of human ZNF205 (aa 503-553). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction