Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF207 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530641 Supplier Invitrogen™ Supplier No. PA530641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Xenopus laevis (81%), Rhesus Monkey (100%), Chimpanzee (83%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZNF207 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF207
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF207
Gene Accession No. O43670
Gene Alias 49 kDa zinc finger protein; 8430401D15Rik; BUB3-interacting and GLEBS motif-containing protein ZNF207; BUGZ; hBuGZ; Zep; Zfp207; Zinc finger protein 207; Znf207
Gene Symbols ZNF207
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF207. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.