Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF207 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564139 Supplier Invitrogen™ Supplier No. PA564139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WCWYCNRDFD DEKILIQHQK AKHFKCHICH KKLYTGPGLA IHCMQVHKET IDAVPNAIPG RTDIELEIYG MEGIPEKDMD ERRRLLEQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ZNF207 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF207
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF207
Gene Accession No. O43670
Gene Alias 49 kDa zinc finger protein; 8430401D15Rik; BUB3-interacting and GLEBS motif-containing protein ZNF207; BUGZ; hBuGZ; Zep; Zfp207; Zinc finger protein 207; Znf207
Gene Symbols ZNF207
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF207. Recombinant protein control fragment (Product #RP-103533).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.