Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF211 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | ZNF211 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4918
![]()
|
Proteintech Group Inc
254581AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
May be involved in transcriptional regulation.Specifications
| ZNF211 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13398 | |
| 10520 | |
| ZNF211 Fusion Protein Ag22135 | |
| Primary | |
| -20°C | |
| ZNF211 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C2H2 25, CH2H2 25, zinc finger protein 211, Zinc finger protein C2H2 25, ZNF 25, ZNF211, ZNFC25 | |
| ZNF211 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title