Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF212 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565846
Description
Immunogen sequence: LPGEFSCIAEE QAFLSPEQTE LWGGQGSSVL LETGPGDSTL EEPVGSRVPS SSRTVGCPKQ KSHRQVQLDQ ECGQGLKLK Highest antigen sequence identity to the following orthologs - mouse 58%, rat 57%.
This gene belongs to the C2H2-type zinc finger gene family. The zinc finger proteins are involved in gene regulation and development, and are quite conserved throughout evolution. Like this gene product, a third of the zinc finger proteins containing C2H2 fingers also contain the KRAB domain, which has been found to be involved in protein-protein interactions.
Specifications
ZNF212 | |
Polyclonal | |
Unconjugated | |
ZNF212 | |
C2H2-150; Zinc finger protein 212; Zinc finger protein C2H2-150; ZNF182; ZNF212; ZNFC150 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7988 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UDV6 | |
ZNF212 | |
Recombinant Human ZNF212. Recombinant protein control fragment (Product #RP-106513). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction