Learn More
ZNF213 Monoclonal Antibody (3D3)

Description
ZNF213 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | ZNF213 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3D3 |
| Concentration | 0.2 to 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF213 |
| Gene Accession No. | O14771 |
| Gene Alias | CR53; putative transcription factor CR53; Zinc finger protein 213; zinc finger protein with KRAB and SCAN domains 21; ZKSCAN21; ZNF213; ZSCAN53 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.