Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF213 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583255 Supplier Invitrogen™ Supplier No. PA583255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAQRDLFWDI KRENSRNTTL GFGLKGQSEK SLLQEMVPVV PGQTGSDVTV SWSPEEAEAW ESENRP Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 51%.

C2H2 zinc finger proteins, such as ZNF213, have bipartite structures in which one domain binds DNA or RNA and the other modulates target gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF213
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF213
Gene Accession No. O14771
Gene Alias CR53; putative transcription factor CR53; Zinc finger protein 213; zinc finger protein with KRAB and SCAN domains 21; ZKSCAN21; ZNF213; ZSCAN53
Gene Symbols ZNF213
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF213. Recombinant protein control fragment (Product #RP-97583).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7760
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.