Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF214 Polyclonal Antibody

ZNF214 Polyclonal Antibody
Supplier: Thermo Scientific PA541136
Description
ZNF214 Polyclonal Antibody for Western Blot

Specifications
ZNF214 | |
Polyclonal | |
Unconjugated | |
ZNF214 | |
BAZ 1; BAZ1; BAZ-1; BWSCR2-associated zinc finger protein 1; zinc finger protein 214; ZNF214 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UL59 | |
ZNF214 | |
Synthetic peptide directed towards the N-terminal of human ZNF214 (aa 72-121). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction