Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF215 Monoclonal Antibody (2C11)
GREENER_CHOICE

Catalog No. PIMA520296
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520296 Supplier Invitrogen™ Supplier No. MA520296
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MQPLSKLMAI SKPRNLSLRE QREVLRADMS WQQETNPVVE THDSEASRQK FRHFQYLKVS GPHEALSQLW ELCLQWLRPE IHTKKQIIEL LVLEQFLAIL PEEVRTWVNL QHPNNSKDMV TLIEDVIEML EDEDMPCKDS ALQMGSIKEK MKAGSRTGKP QEPVTFKDVV VEFSKEEWGQ LDSAVKNLYR NVMLENFRNL NSLRKAHLLS KPFESLKLES KKKRWIMEKE IPRKTIFDMK SISGEESSHG VIMTRLTESG HPSSDAWKDG VSLLLPRLEY NDAVSAHCNL HLWVQVILLP QPPE.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF215
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C11
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF215
Gene Accession No. Q9UL58
Gene Alias BAZ 2; BAZ2; BAZ-2; BWSCR2-associated zinc finger protein 2; zinc finger protein 215; Zinc finger protein with KRAB and SCAN domains 11; ZKSCAN11; ZNF215; ZSCAN43
Gene Symbols ZNF215
Host Species Mouse
Immunogen ZNF215 (AAH14538, 1 a.a. ~ 304 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7762
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.