Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF217 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178188JF646
Description
ZNF217 Polyclonal antibody specifically detects ZNF217 in Human samples. It is validated for Western BlotSpecifications
ZNF217 | |
Polyclonal | |
Western Blot | |
DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518 | |
The immunogen for this product maps to a region between residue 100 and 150 of human Zinc Finger Protein 217 using the numbering given in entry NP_006517.1 (GeneID 7764). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7764 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction