Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF217 Antibody (OTI3G5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274933AF647
Description
ZNF217 Monoclonal specifically detects ZNF217 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF217 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF217 | |
Recombinant protein fragment corresponding to amino acids 730-1048 of human ZNF217 (NP_006517) produced in E.coli. | |
0.1 mL | |
7764.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G5 | |
Western Blot, Immunohistochemistry | |
DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction