Learn More
ZNF217 Mouse anti-Human, Clone: OTI2B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809052

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at ′Ser-473′.Specifications
ZNF217 | |
Monoclonal | |
Unconjugated | |
ZNF217 | |
ZABC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7764 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75362 | |
ZNF217 | |
Human recombit protein fragment corresponding to amino acids 730-1048 of human ZNF217 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.