Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF219 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521120
Description
A suggested positive control is mouse brain tissue lysate. PA5-21120 can be used with blocking peptide PEP-1234.
ZNF219 is a member of the Kruppel-like zinc finger family that is involved in a diverse range of biological processes. ZNF219 is a 77-kDa nuclear protein containing nine sets of C2H2 zinc finger structures. It may be involved in transcriptional regulation, it functions as a transcriptional repressor for the HMGN1 promoter.
Specifications
ZNF219 | |
Polyclonal | |
Unconjugated | |
ZNF219 | |
2010302A17Rik; ZFP219; Zinc finger protein 219; ZNF219 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305848, 51222, 69890 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6IQX8, Q9P2Y4 | |
Zfp219, ZNF219 | |
An 18 amino acid peptide near the amino terminus of human ZFP219. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction