Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF219 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584279
Description
Immunogen sequence: EAEEETWARG RSLGSLASLH PRPGEGPGHS ASAAGAQARS TATQEENGLL VGGTRPEGGR GATGKDCPFC GKSFRSAHHL K.
ZNF219 is a member of the Kruppel-like zinc finger family that is involved in a diverse range of biological processes. ZNF219 is a 77-kDa nuclear protein containing nine sets of C2H2 zinc finger structures. It may be involved in transcriptional regulation, it functions as a transcriptional repressor for the HMGN1 promoter.
Specifications
ZNF219 | |
Polyclonal | |
Unconjugated | |
ZNF219 | |
2010302A17Rik; ZFP219; Zinc finger protein 219; ZNF219 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2Y4 | |
ZNF219 | |
Recombinant protein corresponding to Human ZNF219. Recombinant protein control fragment (Product #RP-101450). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction