Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF219 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111004 Supplier Invitrogen™ Supplier No. PA5111004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THQPERPRSP AARLLLELEE RALLREARLG RARSSGGMQA TPATEGLARP QAPSSSAFRC PYCKGKFRTS AERERHLHIL HRPWKCGLCS FGSSQEEELL HHSLTAHGAP ERPLAATSAA PPPQPQPQPP PQPEPRSVP Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 84%.

ZNF219 is a member of the Kruppel-like zinc finger family that is involved in a diverse range of biological processes. ZNF219 is a 77-kDa nuclear protein containing nine sets of C2H2 zinc finger structures. It may be involved in transcriptional regulation, it functions as a transcriptional repressor for the HMGN1 promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF219
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF219
Gene Accession No. Q9P2Y4
Gene Alias 2010302A17Rik; ZFP219; Zinc finger protein 219; ZNF219
Gene Symbols ZNF219
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZNF219. Recombinant protein control fragment (Product #RP-89136).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.