Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540977 Supplier Invitrogen™ Supplier No. PA540977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TGEKPYQCSE CGKCFSQSSH LRQHMKVHKE EKPRKTRGKN IRVKTHLPSW Sequence homology: Cow: 91%; Dog: 100%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 93%.

ZNF22 encodes a protein that is expressed in the epithelial component of the developing tooth organ during early bud and cap stages as well as in osteoblasts of craniofacial bone and the developing tongue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF22
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF22
Gene Accession No. P17026
Gene Alias 2900028O21Rik; AU041373; HKR-T1; Kox15; Krox25; Krox-25; Krox-25-2; Krox26; Krox-26; krox-26 protein; krueppel-type zinc finger protein Krox-25; Zfp422; Zinc finger protein 22; zinc finger protein 22 (KOX 15); zinc finger protein 422; Zinc finger protein KOX15; zinc finger protein Krox-25; Zinc finger protein Krox-26; ZNF22; ZNF422
Gene Symbols ZNF22
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF22 (aa 162-211).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7570
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.