Learn More
Invitrogen™ ZNF22 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ZNF22 protein.
Specifications
Specifications
| Antigen | ZNF22 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ZNF22 |
| Gene Accession No. | P17026 |
| Gene Alias | 2900028O21Rik; AU041373; HKR-T1; Kox15; Krox25; Krox-25; Krox-25-2; Krox26; Krox-26; krox-26 protein; krueppel-type zinc finger protein Krox-25; Zfp422; Zinc finger protein 22; zinc finger protein 22 (KOX 15); zinc finger protein 422; Zinc finger protein KOX15; zinc finger protein Krox-25; Zinc finger protein Krox-26; ZNF22; ZNF422 |
| Gene Symbols | ZNF22 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.