Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF22 Polyclonal Antibody
ZNF22 Polyclonal Antibody
Supplier: Thermo Scientific PA553481
Description
ZNF22 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
ZNF22 | |
Polyclonal | |
Unconjugated | |
ZNF22 | |
2900028O21Rik; AU041373; HKR-T1; Kox15; Krox25; Krox-25; Krox-25-2; Krox26; Krox-26; krox-26 protein; krueppel-type zinc finger protein Krox-25; Zfp422; Zinc finger protein 22; zinc finger protein 22 (KOX 15); zinc finger protein 422; Zinc finger protein KOX15; zinc finger protein Krox-25; Zinc finger protein Krox-26; ZNF22; ZNF422 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17026 | |
ZNF22 | |
Recombinant protein corresponding to Human ZNF22. Recombinant protein control fragment (Product #RP-91685). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction