Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF223 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541137 Supplier Invitrogen™ Supplier No. PA541137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HEGWSCQQIW EEIASDLTRP QDSTIKSSQF FEQGDAHSQV EEGISIMHTG Sequence homology: Human: 100%.

ZNF223 is part of a ZNF gene cluster located on human chromosome 19q13.2 which encodes Kruppel-associated box (KRAB) motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF223
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF223
Gene Accession No. Q9UK11
Gene Alias Homo sapiens zinc finger protein 223; KRAB A domain; zinc finger protein 223; ZNF223
Gene Symbols ZNF223
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF223.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7766
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.